4-Brom-2-ethylanilin ≥97%

Lieferant: Thermo Scientific
Gefahr

L12725.06 L12725.14
L12725.06EA 49.7 EUR
L12725.06 L12725.14
4-Brom-2-ethylanilin ≥97%
4-Brom-2-ethylanilin

Purity: 97% CAS No.: 45762-41-2 Molecular Formula: C8H10BrN Formula Weight: 200.08 Refractive Index: 1.6020 Boiling Point: 110-112°/2mm UN No.: 2810 MDL No.: MFCD00041434

Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
Siedepunkt: 110…112 °C (2 mmHg)
Dichte: 1,412
Lagertemperatur: Raumtemperatur
MDL: MFCD00041434
CAS-Nummer: 45762-41-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR