4-Brom-2,5-xylidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2,5-dimethylaniline

OT-0070-5G OT-0070-25G OT-0070-1G
COBBOT-0070-5GEA 42.3 EUR
COBBOT-0070-5G COBBOT-0070-25G COBBOT-0070-1G
4-Brom-2,5-xylidin
4-Brom-2,5-xylidin
Formel: C₈H₁₀BrN
Molecular Weight: 200.0757 g/mol
MDL: MFCD07780642
CAS-Nummer: 30273-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR