2,6-Dichlor-m-toluidin

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dichloro-3-methylaniline

AN-1197-5G AN-1197-25G AN-1197-1G
COBBAN-1197-5GEA 69 EUR
COBBAN-1197-5G COBBAN-1197-25G COBBAN-1197-1G
2,6-Dichlor-m-toluidin
2,6-Dichlor-m-toluidin
Formel: C₇H₇Cl₂N
Molecular Weight: 176,0432 g/mol
MDL: MFCD00007676
CAS-Nummer: 64063-37-2
EINECS: 264-649-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR