2-Fluor-3-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-3-(trifluoromethyl)benzeneboronic acid
BB-3557-25G
BB-3557-1G
BB-3557-5G
BB-3557-100G
COBBBB-3557-25GEA
0
EUR
COBBBB-3557-25G
COBBBB-3557-1G
COBBBB-3557-5G
COBBBB-3557-100G
2-Fluor-3-(trifluormethyl)phenylboronsäure
2-Fluor-3-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ |
MDL:
MFCD07784336 CAS-Nummer: 157834-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...