2-Fluor-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-(trifluoromethyl)benzeneboronic acid

BB-2788-5G BB-2788-25G BB-2788-1G
COBBBB-2788-5GEA 0 EUR
COBBBB-2788-5G COBBBB-2788-25G COBBBB-2788-1G
2-Fluor-5-(trifluormethyl)phenylboronsäure
2-Fluor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Molecular Weight: 207.92 g/mol
MDL: MFCD03701538
CAS-Nummer: 352535-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR