2-Fluor-5-(trifluormethyl)phenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-5-(trifluoromethyl)benzeneboronic acid
016257-1G
016257-5G
FLUO016257-1GEA
31.1
EUR
FLUO016257-1G
FLUO016257-5G
2-Fluor-5-(trifluormethyl)phenylboronsäure
2-Fluor-5-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.92 g/mol |
MDL:
MFCD03701538 CAS-Nummer: 352535-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...