2-Fluor-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid

BB-3361-1G BB-3361-5G
COBBBB-3361-1GEA 26.2 EUR
COBBBB-3361-1G COBBBB-3361-5G
2-Fluor-5-methylphenylboronsäure
2-Fluor-5-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,9466 g/mol
MDL: MFCD03428512
CAS-Nummer: 166328-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR