3-Fluor-2-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Fluoro-2-(trifluoromethyl)phenyl)boronic acid, 2-(Trifluoromethyl)-3-fluoro-phenylboronic acid
FA-1581-1G
FA-1581-250M
COBBFA-1581-1GEA
0
EUR
COBBFA-1581-1G
COBBFA-1581-250M
3-Fluor-2-(trifluormethyl)phenylboronsäure
3-Fluor-2-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.918 g/mol |
MDL:
MFCD08706086 CAS-Nummer: 850411-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...