3-Fluor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-2-methylphenyl)boronic acid, 3-Fluoro-2-methylbenzeneboronic acid

BB-3475-5G BB-3475-1G
COBBBB-3475-5GEA 362 EUR
COBBBB-3475-5G COBBBB-3475-1G
3-Fluor-2-methylphenylboronsäure
3-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD07368679
CAS-Nummer: 163517-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR