3-Fluor-4-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Fluoro-4-(trifluoromethyl)phenyl)boronic acid, 3-Fluoro-4-trifluoromethylphenylboronic acid
BB-8824-5G
BB-8824-1G
COBBBB-8824-5GEA
571
EUR
COBBBB-8824-5G
COBBBB-8824-1G
3-Fluor-4-(trifluormethyl)phenylboronsäure
3-Fluor-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.92 g/mol |
MDL:
MFCD08706261 CAS-Nummer: 864759-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...