3-Fluor-4-methylphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Fluoro-4-methylbenzeneboronic acid

B24512.06 B24512.03 B24512.14
B24512.06EA 91.8 EUR
B24512.06 B24512.03 B24512.14
3-Fluor-4-methylphenylboronsäure ≥98%
3-Fluor-4-methylphenylboronsäure

Purity: 98% CAS No.: 168267-99-0 Molecular Formula: C7H8BFO2 Formula Weight: 153.95 Melting Point: 234° Beilstein Reference: 8544594

Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683115
CAS-Nummer: 168267-99-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR