3-Fluor-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-4-methylbenzeneboronic acid

BB-2209-5G BB-2209-25G BB-2209-1G
COBBBB-2209-5GEA 50.9 EUR
COBBBB-2209-5G COBBBB-2209-25G COBBBB-2209-1G
3-Fluor-4-methylphenylboronsäure
3-Fluor-4-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD02683115
CAS-Nummer: 168267-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR