3-Fluor-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluoro-5-(trifluoromethyl)benzeneboronic acid, (3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid, 3-(Trifluoromethyl)-5-fluorophenylboronic acid
BB-3488-5G
BB-3488-1G
COBBBB-3488-5GEA
0
EUR
COBBBB-3488-5G
COBBBB-3488-1G
3-Fluor-5-(trifluormethyl)phenylboronsäure
3-Fluor-5-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ |
MDL:
MFCD06657735 CAS-Nummer: 159020-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...