3-Fluor-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-5-methylphenyl)boronic acid, 3-Fluoro-5-methylbenzeneboronic acid

BB-2784-5G BB-2784-1G
COBBBB-2784-5GEA 303 EUR
COBBBB-2784-5G COBBBB-2784-1G
3-Fluor-5-methylphenylboronsäure
3-Fluor-5-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
MDL: MFCD07363785
CAS-Nummer: 850593-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR