5-Fluor-2-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-9048-1G
BB-9048-5G
COBBBB-9048-1GEA
0
EUR
COBBBB-9048-1G
COBBBB-9048-5G
5-Fluor-2-(trifluormethyl)phenylboronsäure
5-Fluor-2-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.9180 g/mol |
MDL:
MFCD06656275 CAS-Nummer: 928053-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...