5-Fluor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

BB-2627-25G BB-2627-5G BB-2627-1G
COBBBB-2627-25GEA 153 EUR
COBBBB-2627-25G COBBBB-2627-5G COBBBB-2627-1G
5-Fluor-2-methylphenylboronsäure
5-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD03095047
CAS-Nummer: 163517-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR