5-Fluor-2-methylphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

016255-1G 016255-25G 016255-5G
FLUO016255-1GEA 31.1 EUR
FLUO016255-1G FLUO016255-25G FLUO016255-5G
5-Fluor-2-methylphenylboronsäure
5-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD03095047
CAS-Nummer: 163517-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR