4-Fluor-3-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid
BB-2793-5G
BB-2793-25G
BB-2793-1G
COBBBB-2793-5GEA
86.3
EUR
COBBBB-2793-5G
COBBBB-2793-25G
COBBBB-2793-1G
4-Fluor-3-(trifluormethyl)phenylboronsäure
4-Fluor-3-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ |
MDL:
MFCD07363787 CAS-Nummer: 182344-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...