4-Fluor-3-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid
BB-2127-25G
BB-2127-100G
BB-2127-5G
COBBBB-2127-25GEA
127
EUR
COBBBB-2127-25G
COBBBB-2127-100G
COBBBB-2127-5G
4-Fluor-3-methylphenylboronsäure
4-Fluor-3-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153,95 g/mol |
MDL:
MFCD01863527 CAS-Nummer: 139911-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...