2'-Fluor-3'-(trifluormethyl)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone
006373-1G
006373-5G
FLUO006373-1GEA
40.1
EUR
FLUO006373-1G
FLUO006373-5G
2'-Fluor-3'-(trifluormethyl)acetophenon
2'-Fluor-3'-(trifluormethyl)acetophenon
Formel:
C₉H₆F₄O |
MDL:
MFCD00061145 CAS-Nummer: 207853-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...