2'-Fluor-4'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Fluoro-4-(trifluoromethyl)phenyl)ethanone

006374-1G 006374-5G
FLUO006374-1GEA 72.9 EUR
FLUO006374-1G FLUO006374-5G
2'-Fluor-4'-(trifluormethyl)acetophenon
2'-Fluor-4'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
MDL: MFCD00061247
CAS-Nummer: 122023-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR