2'-Fluor-6'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

006376-1G 006376-5G
FLUO006376-1GEA 45.9 EUR
FLUO006376-1G FLUO006376-5G
2'-Fluor-6'-(trifluormethyl)acetophenon
2'-Fluor-6'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206,14 g/mol
MDL: MFCD00061249
CAS-Nummer: 174013-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR