3'-Fluor-4'-(trifluormethyl)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanone
007905-5G
007905-1G
FLUO007905-5GEA
231
EUR
FLUO007905-5G
FLUO007905-1G
3'-Fluor-4'-(trifluormethyl)acetophenon
3'-Fluor-4'-(trifluormethyl)acetophenon
Formel:
C₉H₆F₄O |
MDL:
MFCD00236273 CAS-Nummer: 237761-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...