3'-Fluor-5'-(trifluormethyl)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-[3-Fluoro-5-(trifluoromethyl)phenyl]ethan-1-one, 3-Acetyl-5-fluorobenzotrifluoride
006377-1G
006377-5G
FLUO006377-1GEA
36.9
EUR
FLUO006377-1G
FLUO006377-5G
3'-Fluor-5'-(trifluormethyl)acetophenon
3'-Fluor-5'-(trifluormethyl)acetophenon
Formel:
C₉H₆F₄O |
MDL:
MFCD00061250 CAS-Nummer: 202664-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...