4'-Fluor-2'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Fluoro-2-(trifluoromethyl)phenyl)ethanone

006378-1G 006378-5G
FLUO006378-1GEA 31.1 EUR
FLUO006378-1G FLUO006378-5G
4'-Fluor-2'-(trifluormethyl)acetophenon
4'-Fluor-2'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
MDL: MFCD00061251
CAS-Nummer: 208173-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR