2,3-Difluor-4-methylbenzylamin

Lieferant: FLUOROCHEM

005756-1G 005756-5G
FLUO005756-1GEA 0 EUR
FLUO005756-1G FLUO005756-5G
2,3-Difluor-4-methylbenzylamin
2,3-Difluor-4-methylbenzylamin
Formel: C₈H₉F₂N
MDL: MFCD01631409
CAS-Nummer: 261763-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR