5-Fluor-2-methylbenzylamin
Lieferant: FLUOROCHEM
Synonyme:
(5-Fluoro-2-methylphenyl)methanamine
005798-1G
005798-5G
005798-10G
FLUO005798-1GEA
60.8
EUR
FLUO005798-1G
FLUO005798-5G
FLUO005798-10G
5-Fluor-2-methylbenzylamin
5-Fluor-2-methylbenzylamin
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol |
MDL:
MFCD01631526 CAS-Nummer: 261951-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...