2-(Difluormethoxy)benzylamin

Lieferant: FLUOROCHEM

007858-5G 007858-1G
FLUO007858-5GEA 210 EUR
FLUO007858-5G FLUO007858-1G
2-(Difluormethoxy)benzylamin
2-(Difluormethoxy)benzylamin
Formel: C₈H₉F₂NO
MDL: MFCD00236229
CAS-Nummer: 243863-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR