3-Fluor-4-methoxybenzylamin

Lieferant: CHEMPUR

Synonyme: (3-Fluoro-4-methoxyphenyl)methanamine

FL13341.POR 013341-5G 013341-1G
CHMPFL13341.POREA 0 EUR
CHMPFL13341.POR FLUO013341-5G FLUO013341-1G
3-Fluor-4-methoxybenzylamin
3-Fluor-4-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
MDL: MFCD04116360
CAS-Nummer: 123652-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR