2-Hydroxyethyl-phenylsulfid

Lieferant: FLUOROCHEM

Synonyme: 2-(Phenylthio)ethanol

003534-100G 003534-25G
FLUO003534-100GEA 252 EUR
FLUO003534-100G FLUO003534-25G
2-Hydroxyethyl-phenylsulfid
2-Hydroxyethyl-phenylsulfid
Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
MDL: MFCD00002905
CAS-Nummer: 699-12-7
EINECS: 211-828-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR