2-Brom-2'-chloracetanilid

Lieferant: Thermo Scientific

Synonyme: N-(Bromoacetyl)-2-chloroaniline

SPB05477DA SPB05477EA
MAYBSPB05477DAEA 0 EUR
MAYBSPB05477DA MAYBSPB05477EA
2-Brom-2'-chloracetanilid
2-Brom-2'-chloracetanilid
Formel: C₈H₇BrClNO
MDL: MFCD00173815
CAS-Nummer: 5439-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR