2'-Brom-4'-chloracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2-Bromo-4-chlorophenyl)ethanone
011300-1G
011300-5G
FLUO011300-1GEA
91.1
EUR
FLUO011300-1G
FLUO011300-5G
2'-Brom-4'-chloracetophenon
2'-Brom-4'-chloracetophenon
Formel:
C₈H₆BrClO Molecular Weight: 233,4896 g/mol |
MDL:
MFCD02683063 CAS-Nummer: 825-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...