5'-Brom-2'-methoxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(5-Bromo-2-methoxyphenyl)ethanone

019294-5G 019294-25G
FLUO019294-5GEA 210 EUR
FLUO019294-5G FLUO019294-25G
5'-Brom-2'-methoxyacetophenon
5'-Brom-2'-methoxyacetophenon
Formel: C₉H₉BrO₂
Molecular Weight: 229.0706 g/mol
MDL: MFCD03789141
CAS-Nummer: 16740-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR