2-Brom-2',4'-difluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-1-(2,4-difluorophenyl)ethanone, 2,4-Difluorophenacyl bromide

M-Clarity™ Qualitätsstufe = MQ100

595152-1G 595152-5G
SIAL595152-1GEA 29.8 EUR
SIAL595152-1G SIAL595152-5G
2-Brom-2',4'-difluoracetophenon, Sigma-Aldrich®
2-Brom-2',4'-difluoracetophenon
Formel: C₈H₅BrF₂O
Molecular Weight: 235,03 g/mol
MDL: MFCD00142822
CAS-Nummer: 102429-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR