3'-Brom-2,2,2-trifluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Bromophenyl)-2,2,2-trifluoroethanone

M-Clarity™ Qualitätsstufe = MQ100

689238-1G 689238-5G
SIAL689238-1GEA 49.7 EUR
SIAL689238-1G SIAL689238-5G
3'-Brom-2,2,2-trifluoracetophenon, Sigma-Aldrich®
3'-Brom-2,2,2-trifluoracetophenon
Formel: C₈H₄BrF₃O
Molecular Weight: 253.0160 g/mol
MDL: MFCD00511232
CAS-Nummer: 655-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR