Dibutylzinnbis(2-ethylhexanoat)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Di-n-butyltin bis(2-ethylhexanoate), Dibutylstannanediyl bis(2-ethylhexanoate)

71126.06 71126.14 71126.22
71126.06EA 36.1 EUR
71126.06 71126.14 71126.22
Dibutylzinnbis(2-ethylhexanoat)
Dibutylzinnbis(2-ethylhexanoat)

CAS No.: 2781-10-4 Molecular Formula: C24H48O4Sn Formula Weight: 519.34 Physical Form: Lump Density: 1.070 Odour: Odourless Boiling Point: 220°/2mm Solubility: Soluble in mineral oil, acetone, xylene UN No.: 3146 MDL No.: MFCD00015261

Formel: C₂₄H₄₈O₄Sn
Molecular Weight: 519,34 g/mol
Siedepunkt: 220 °C (2 mmHg)
Schmelzpunkt: 57…59 °C
Dichte: 1,070
Lagertemperatur: Raumtemperatur
MDL: MFCD00015261
CAS-Nummer: 2781-10-4
EINECS: 220-481-2
UN: 3146
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR