3',5'-Difluor-2'-hydroxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Difluoro-2-hydroxyphenyl)ethanone

004816-1G 004816-5G
FLUO004816-1GEA 73.1 EUR
FLUO004816-1G FLUO004816-5G
3',5'-Difluor-2'-hydroxyacetophenon
3',5'-Difluor-2'-hydroxyacetophenon
Formel: C₈H₆F₂O₂
MDL: MFCD00042476
CAS-Nummer: 140675-42-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR