2-Chlor-6-fluor-3-methylbenzylamin

Lieferant: FLUOROCHEM

005652-1G 005652-5G
FLUO005652-1GEA 282 EUR
FLUO005652-1G FLUO005652-5G
2-Chlor-6-fluor-3-methylbenzylamin
2-Chlor-6-fluor-3-methylbenzylamin
Formel: C₈H₉ClFN
Molecular Weight: 173.62 g/mol
MDL: MFCD01631395
CAS-Nummer: 261762-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR