6-Chlor-2-fluor-3-methylbenzylamin

Lieferant: FLUOROCHEM

005653-1G 005653-5G
FLUO005653-1GEA 0 EUR
FLUO005653-1G FLUO005653-5G
6-Chlor-2-fluor-3-methylbenzylamin
6-Chlor-2-fluor-3-methylbenzylamin
Formel: C₈H₉ClFN
MDL: MFCD01631420
CAS-Nummer: 261762-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR