4'-Fluor-2'-methoxyacetophenon
Lieferant: FLUOROCHEM
006262-1G
006262-5G
FLUO006262-1GEA
31.1
EUR
FLUO006262-1G
FLUO006262-5G
4'-Fluor-2'-methoxyacetophenon
4'-Fluor-2'-methoxyacetophenon
Formel:
C₉H₉FO₂ |
MDL:
MFCD00061143 CAS-Nummer: 51788-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...