4'-Fluor-3'-methoxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Fluoro-3-methoxyphenyl)ethanone

013010-1G 013010-5G
FLUO013010-1GEA 107 EUR
FLUO013010-1G FLUO013010-5G
4'-Fluor-3'-methoxyacetophenon
4'-Fluor-3'-methoxyacetophenon
Formel: C₉H₉FO₂
Molecular Weight: 168.17 g/mol
MDL: MFCD00272135
CAS-Nummer: 64287-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR