2'-Methoxy-2,2,2-trifluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
2,2,2-Trifluoro-1-(2-methoxyphenyl)ethanone
023147-1G
023147-5G
FLUO023147-1GEA
61.1
EUR
FLUO023147-1G
FLUO023147-5G
2'-Methoxy-2,2,2-trifluoracetophenon
2'-Methoxy-2,2,2-trifluoracetophenon
Formel:
C₉H₇F₃O₂ Molecular Weight: 204.15 g/mol |
MDL:
MFCD00461894 CAS-Nummer: 26944-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...