4-Brom-2,5-xylenol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2,5-dimethylphenol

OT-0391-5G OT-0391-25G OT-0391-1G
COBBOT-0391-5GEA 471 EUR
COBBOT-0391-5G COBBOT-0391-25G COBBOT-0391-1G
4-Brom-2,5-xylenol
4-Brom-2,5-xylenol
Formel: C₈H₉BrO
Molecular Weight: 201,0600 g/mol
MDL: MFCD00029746
CAS-Nummer: 85223-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR