2-Chlor-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3834-5G BB-3834-25G BB-3834-1G
COBBBB-3834-5GEA 310 EUR
COBBBB-3834-5G COBBBB-3834-25G COBBBB-3834-1G
2-Chlor-3-(trifluormethyl)phenylboronsäure
2-Chlor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
Molecular Weight: 224.3726 g/mol
MDL: MFCD09258745
CAS-Nummer: 957061-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR