2-Chlor-6-fluor-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8431-1G BB-8431-250M
COBBBB-8431-1GEA 143 EUR
COBBBB-8431-1G COBBBB-8431-250M
2-Chlor-6-fluor-5-methylphenylboronsäure
2-Chlor-6-fluor-5-methylphenylboronsäure
Formel: C₇H₇BClFO₂
Molecular Weight: 188.3917 g/mol
MDL: MFCD03701535
CAS-Nummer: 352535-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR