3-Chlor-4-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-4-(trifluoromethyl)benzeneboronic acid

BB-2644-5G BB-2644-25G BB-2644-1G
COBBBB-2644-5GEA 385 EUR
COBBBB-2644-5G COBBBB-2644-25G COBBBB-2644-1G
3-Chlor-4-(trifluormethyl)phenylboronsäure
3-Chlor-4-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
Molecular Weight: 224,38 g/mol
MDL: MFCD03788417
CAS-Nummer: 847756-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR