Tetrahydro-4H-thiopyran-4-on
Lieferant: COMBI-BLOCKS
Synonyme:
Tetrahydrothiopyran-4-one
OS-7587-5G
OS-7587-25G
OS-7587-1G
COBBOS-7587-5GEA
30.8
EUR
COBBOS-7587-5G
COBBOS-7587-25G
COBBOS-7587-1G
Tetrahydro-4H-thiopyran-4-on
Tetrahydro-4H-thiopyran-4-on
Formel:
C₅H₈OS Molecular Weight: 116,18 g/mol |
MDL:
MFCD00006660 CAS-Nummer: 1072-72-6 EINECS: 214-015-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...