3',5'-Dibrom-4'-hydroxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Dibromo-4-hydroxyphenyl)ethanone

009646-10G 009646-25G 009646-5G
FLUO009646-10GEA 0 EUR
FLUO009646-10G FLUO009646-25G FLUO009646-5G
3',5'-Dibrom-4'-hydroxyacetophenon
3',5'-Dibrom-4'-hydroxyacetophenon
Formel: C₈H₆Br₂O₂
MDL: MFCD00075779
CAS-Nummer: 2887-72-1
EINECS: 220-750-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR