3',5'-Dibrom-2'-hydroxyacetophenon 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(3,5-Dibromo-2-hydroxyphenyl)ethanone
B22413.06
B22413.14
B22413.06EA
36
EUR
B22413.06
B22413.14
3',5'-Dibrom-2'-hydroxyacetophenon 99%
3',5'-Dibrom-2'-hydroxyacetophenon
Purity: 99%
CAS No.: 22362-66-9
Molecular Formula: C8H6Br2O2
Formula Weight: 293.95
Melting Point: 108-111°
MDL No.: MFCD00191992
Formel:
C₈H₆Br₂O₂ Molecular Weight: 293,95 g/mol Schmelzpunkt: 108…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191992 CAS-Nummer: 22362-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...