2'-Hydroxy-5'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Hydroxy-5-nitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

646393-10G 646393-1G
SIAL646393-10GEA 66.9 EUR
SIAL646393-10G SIAL646393-1G
2'-Hydroxy-5'-nitroacetophenon, Sigma-Aldrich®
2'-Hydroxy-5'-nitroacetophenon
Formel: C₈H₇NO₄
Molecular Weight: 181,1500 g/mol
MDL: MFCD00463816
CAS-Nummer: 1450-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR